accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16561-29-8

IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
Synonyms: phorbol ester tpa (phorbol derivative) tpa (tetradecanoylphorbol acetate) PMA tetradecanoylphorbol acetate, 4a α isomer factor A1 4a α isomer tetradecanoylphorbol acetate myristate acetate, phorbol acetate, tetradecanoylphorbol phorbol-myristate acetate
Molecular Weight (g/mol): 616.84
Molecular Formula: C36H56O8
InChi Key: PHEDXBVPIONUQT-RGYGYFBISA-N
SMILES: CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)