accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOCH3OHNH2

CAS RN 16655-44-0

IUPAC Name: diphenylboranyl 2-amino-3-hydroxybutanoate
Synonyms: diphenylboranyl 2-amino-3-hydroxybutanoate
Molecular Weight (g/mol): 283.13
Molecular Formula: C16H18BNO3
InChi Key: YWSHJDZMGRKKKP-UHFFFAOYNA-N
SMILES: CC(O)C(N)C(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results