accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 1674-38-0

IUPAC Name: 1-phenyldodecan-1-one
Synonyms: 1-dodecanone, 1-phenyl-
Molecular Weight (g/mol): 260.42
Molecular Formula: C18H28O
InChi Key: DJNJZIFFCJTUDS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results