CAS RN 1684398-38-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mono-2-ethylHydroxyhexyl Terephthalate (MEHHTP), TRC
CAS: 1684398-38-6 Molecular Formula: C16H22O5 Molecular Weight (g/mol): 294.34 Synonym: 1,4-Benzenedicarboxylic Acid 1-(2-Ethyl-5-hydroxyhexyl) Ester,MEHHTP; IUPAC Name: 4-(2-ethyl-5-hydroxy-hexoxy)carbonylbenzoic acid SMILES: CCC(CCC(C)O)COC(=O)c1ccc(cc1)C(=O)O