accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3OONHNOH

CAS RN 169280-83-5

IUPAC Name: tert-butyl N-[5-(hydroxymethyl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-[5-(hydroxymethyl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 224.26
Molecular Formula: C11H16N2O3
InChi Key: BORDVYKLAFQRSP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=NC=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results