accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O2H2H2H2H2H2H2H2H

CAS RN 1693-74-9

IUPAC Name: (²H₈)oxolane
Synonyms: deuterated thf
Molecular Weight (g/mol): 80.16
Molecular Formula: C4H8O
InChi Key: WYURNTSHIVDZCO-SVYQBANQSA-N
SMILES: [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)