accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 171364-81-1

CH3OBOH3CH3CH3CCH3O

CAS RN 171364-81-1

IUPAC Name: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Synonyms: 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 246.11
Molecular Formula: C14H19BO3
InChi Key: BATKIZWNRQGSKE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results