CAS RN 17249-77-3
IUPAC Name:
2,3,5-trichlorothiophene
Synonyms:
2,3,5-trichlorothiophene
Molecular Weight (g/mol):
187.46
Molecular Formula:
C4HCl3S
InChi Key:
AZIFHMYTYFPVMV-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(Cl)S1
1
–
1
of
1
results
1
2,3,5-Trichlorothiophene, 97%, Thermo Scientific™
CAS: 17249-77-3 Molecular Formula: C4HCl3S Molecular Weight (g/mol): 187.462 MDL Number: MFCD01318176 InChI Key: AZIFHMYTYFPVMV-UHFFFAOYSA-N Synonym: thiophene, 2,3,5-trichloro,2,3,5-trichloro-thiophene,acmc-1ch2b,thiophene,2,3,5-trichloro PubChem CID: 123374 IUPAC Name: 2,3,5-trichlorothiophene SMILES: C1=C(SC(=C1Cl)Cl)Cl