CAS RN 1738-36-9
IUPAC Name:
2-methoxyacetonitrile
Synonyms:
acetonitrile, methoxy-
Molecular Weight (g/mol):
71.08
Molecular Formula:
C3H5NO
InChi Key:
QKPVEISEHYYHRH-UHFFFAOYSA-N
SMILES:
COCC#N
1
–
5
of
5
results
1
Methoxyacetonitrile, 98%
CAS: 1738-36-9 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00001892 InChI Key: QKPVEISEHYYHRH-UHFFFAOYSA-N PubChem CID: 74442 IUPAC Name: 2-methoxyacetonitrile SMILES: COCC#N
2
2-Methoxyacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Methoxyacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-Methoxyacetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.