accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFFF

CAS RN 17408-14-9

IUPAC Name: 1-[2-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: o-(trifluoromethyl)benzoyl
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: FYDUUODXZQITBF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1C(F)(F)F
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)