accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1749-68-4

CH3NNH2ClN

CAS RN 1749-68-4

IUPAC Name: 6-chloro-2-methylpyrimidin-4-amine
Synonyms: 6-chloro-2-methylpyrimidin-4-amine
Molecular Weight (g/mol): 143.57
Molecular Formula: C5H6ClN3
InChi Key: SSAYHQQUDKQNAC-UHFFFAOYSA-N
SMILES: CC1=NC(N)=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results