accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175278-02-1

H3COOOFFFFFF

CAS RN 175278-02-1

IUPAC Name: ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate
Synonyms: ethyl 3-[3,5-bis(trifluoromethyl)phenyl]-3-oxopropanoate
Molecular Weight (g/mol): 328.21
Molecular Formula: C13H10F6O3
InChi Key: IBRYPSPFMRZTCX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results