accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 175278-33-8

IUPAC Name: 1-bromo-2-(difluoromethoxy)benzene
Synonyms: 1-bromo-2-(difluoromethoxy)benzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: VHEICCMNYWNFQX-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=CC=C1Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results