missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-(difluoromethoxy)benzene, 97+%, Thermo Scientific™

Product Code. 11430767
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430767 10 g 10g
11420767 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430767 Supplier Thermo Scientific Alfa Aesar Supplier No. B23174.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-33-8
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD00221468
InChI Key VHEICCMNYWNFQX-UHFFFAOYSA-N
Synonym 1-bromo-2-difluoromethoxy benzene, 2-difluoromethoxy bromobenzene, benzene, 1-bromo-2-difluoromethoxy, 2-bromophenyl difluoromethyl ether, 2-bromo-alpha,alpha-difluoroanisole, 1-difluoromethoxy-2-bromobenzene, acmc-209e9t, ksc493k0n, 1-bromo-2-difluoromethoxy-benzene, attercop-chm at123241
PubChem CID 2737004
IUPAC Name 1-bromo-2-(difluoromethoxy)benzene
SMILES C1=CC=C(C(=C1)OC(F)F)Br

Specifications

Boiling Point 85°C to 87°C (15 mmHg)
Quantity 10 g
Formula Weight 223.02
Percent Purity ≥97%
Chemical Name or Material 1-Bromo-2-(difluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.