CAS RN 17636-10-1
IUPAC Name:
sodium 3-sulfanylpropane-1-sulfonate
Synonyms:
sodium 3-sulfanylpropane-1-sulfonate
Molecular Weight (g/mol):
178.2
Molecular Formula:
C3H7NaO3S2
InChi Key:
FRTIVUOKBXDGPD-UHFFFAOYSA-M
SMILES:
[Na+].[O-]S(=O)(=O)CCCS
1
–
1
of
1
results
1
Sodium 3-Mercapto-1-propanesulfonate (>90%), TRC
CAS: 17636-10-1 Molecular Formula: C3H7NaO3S2 Molecular Weight (g/mol): 178.21 Synonym: 3-Mercapto-1-propanesulfonic Acid monosodium Salt,3-Mercaptopropanesulfonic Acid, Sodium Salt,Asta 7100,MPS,Sodium 3-mercapto-1-propanesulfonate,Sodium 3-mercaptopropanesulfonate,Sodium 3-mercaptopropylsulfonate,Sodium gamma-mercaptopropanesulfonate IUPAC Name: sodium;3-sulfanylpropane-1-sulfonate SMILES: [Na+].[O-]S(=O)(=O)CCCS