CAS RN 177036-78-1
CAS RN 177036-78-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Alpha-Hydroxy-Beta-methoxy-Beta-phenyl-benzenepropanoic Acid Methyl Ester, TRC
CAS: 177036-78-1 Molecular Formula: C17H18O4 Molecular Weight (g/mol): 286.32 Synonym: (S)-2-Hydroxy-3-methoxy-3,3-diphenylpropionic Acid Methyl Ester,Ambrisentan Hydroxyester Impurity IUPAC Name: methyl (S)-2-hydroxy-3-methoxy-3,3-diphenylpropanoate SMILES: CoC(=O)[C@@H](O)C(OC)(C1=CC=CC=C1)C2=CC=CC=C2