CAS RN 177480-81-8
IUPAC Name:
4-bromo-2-chloro-5-fluorobenzoic acid
Synonyms:
4-bromo-2-chloro-5-fluorobenzoic acid
Molecular Weight (g/mol):
253.45
Molecular Formula:
C7H3BrClFO2
InChi Key:
TZFKKLGCVKORLS-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(F)=C(Br)C=C1Cl
1
–
1
of
1
results
1
4-Bromo-2-chloro-5-fluorobenzoic acid, 98%, Thermo Scientific™
CAS: 177480-81-8 Molecular Formula: C7H3BrClFO2 Molecular Weight (g/mol): 253.45 MDL Number: MFCD00236589 InChI Key: TZFKKLGCVKORLS-UHFFFAOYSA-N Synonym: 4-bromo-2-chloro-5-fluoro-benzoic acid,benzoic acid, 4-bromo-2-chloro-5-fluoro,pubchem1361,acmc-1c1y0,4-bromo-2-chloro-5-fluorobenzoicacid,2-chloro-4-bromo-5-fluorobenzoic acid PubChem CID: 2736225 IUPAC Name: 4-bromo-2-chloro-5-fluorobenzoic acid SMILES: OC(=O)C1=CC(F)=C(Br)C=C1Cl