accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHFOFFF

CAS RN 177596-38-2

IUPAC Name: 3-fluoro-4-(trifluoromethoxy)phenol
Synonyms: 3-fluoro-4-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 196.1
Molecular Formula: C7H4F4O2
InChi Key: UTFSPWRTXCDUIJ-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results