accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FOCH3

CAS RN 177942-47-1

IUPAC Name: 1-(3-fluoro-2-methylphenyl)ethan-1-one
Synonyms: 1-(3-fluoro-2-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: XVVKIGRXVZJHKA-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results