accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHHO

CAS RN 1792-81-0

IUPAC Name: cyclohexane-1,2-diol
Synonyms: 1,2-cyclohexanediol 1,2-cyclohexanediol-trans 1,2-cyclohexanediol-cis
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: PFURGBBHAOXLIO-UHFFFAOYNA-N
SMILES: OC1CCCCC1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results