accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 17994-25-1

HOOO

CAS RN 17994-25-1

IUPAC Name: 1-hydroxycyclopropane-1-carboxylate
Synonyms: 1-hydroxycyclopropane-1-carboxylate
Molecular Weight (g/mol): 101.08
Molecular Formula: C4H5O3
InChi Key: GQXURJDNDYACGE-UHFFFAOYSA-M
SMILES: OC1(CC1)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results