accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 181657-56-7

OH2N

CAS RN 181657-56-7

IUPAC Name: 2-(benzyloxy)cyclopentan-1-amine
Synonyms: 2-(benzyloxy)cyclopentan-1-amine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: JIMSXLUBRRQALI-UHFFFAOYNA-N
SMILES: NC1CCCC1OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results