missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-2-Benzyloxycyclopentylamine, ChiPros 99+%, ee 98%

Product Code. 11343906
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11343906 5 g 5g
11333906 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11343906

Brand: Thermo Scientific Alfa Aesar L17017.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 181657-56-7 | C12H17NO | 191.274 g/mol

(1R,2R)-(-)-2-Benzyloxycyclopentylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(-)-2-Benzyloxycyclopentylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Not miscible or difficult to mix with water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181657-56-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD01075750
InChI Key JIMSXLUBRRQALI-VXGBXAGGSA-N
Synonym 1r,2r-2-benzyloxy cyclopentanamine, 1r,2r-1-amino-2-benzyloxycyclopentane, 1r,2r---2-benzyloxycyclopentylamine, 1r,2r-2-benzyloxy cyclopentan-1-amine, 1r,2r-2-benzyloxycyclopentylamine, 1r,2r-trans-2-benzyloxy-cyclopentylamine, 1r,2r-2-phenylmethoxy cyclopentylamine, 1r,2r-2-benzyloxycyclopentanamine, 1r,2r-2-benzyloxy-cyclopentylamine, 1r-trans-2-phenylmethoxy cyclopentanamine
PubChem CID 11863649
IUPAC Name (1R,2R)-2-phenylmethoxycyclopentan-1-amine
SMILES C1CC(C(C1)OCC2=CC=CC=C2)N

Specifications

Melting Point 23°C
Density 0.983
Boiling Point 90°C to 93°C (0.23 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5305
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Optical Rotation −57.5° (Neat)
Formula Weight 191.27
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclopentylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.