accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18323-96-1

YbH3CCH3H3COHCOFFFFFFFH3CCH3H3COHCOFFFFFFFH3CCH3H3COHCOFFFFFFF

CAS RN 18323-96-1

IUPAC Name: ytterbium(3+) tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-dioxooctan-4-ide)
Synonyms: ytterbium(3+) tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-dioxooctan-4-ide)
Molecular Weight (g/mol): 1058.58
Molecular Formula: C30H30F21O6Yb
InChi Key: QYXREIJIWYJRCU-UHFFFAOYSA-N
SMILES: [Yb+3].CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results