accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 18349-11-6

IUPAC Name: 1-chloro-5-fluoro-4-methyl-2-nitrobenzene
Synonyms: 1-chloro-5-fluoro-4-methyl-2-nitrobenzene
Molecular Weight (g/mol): 189.57
Molecular Formula: C7H5ClFNO2
InChi Key: SJDPAVRCQNFVDM-UHFFFAOYSA-N
SMILES: CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results