Learn More
4-Chloro-2-fluoro-5-nitrotoluene, 98%, Thermo Scientific™
Product Code. 15448428
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448428 25 g 25g
15438428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15448428 Supplier Thermo Scientific Alfa Aesar Supplier No. H61841.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18349-11-6
Molecular Formula C7H5ClFNO2
Molecular Weight (g/mol) 189.57
MDL Number MFCD00134231
InChI Key SJDPAVRCQNFVDM-UHFFFAOYSA-N
PubChem CID 87593
IUPAC Name 1-chloro-5-fluoro-4-methyl-2-nitrobenzene
SMILES CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O
Specifications
Melting Point 42°C to 45°C
Flash Point 113°C (235°F)
Quantity 25 g
Synonym 4-chloro-2-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-4-methyl-2-nitro-benzene, toluene, 4-chloro-2-fluoro-5-nitro, benzene,1-chloro-5-fluoro-4-methyl-2-nitro, benzene, 1-chloro-5-fluoro-4-methyl-2-nitro, acmc-1c7av, 4-chloro-2-fluoro-5-nitro-toluene, buttpark 146\15-31, 2-chloro-4-floro-5-methylnitrobenzene, 2-chloro-4-fluoro-5-methylnitrobenzene
Formula Weight 189.57
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-5-nitrotoluene