accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCl

CAS RN 18368-76-8

IUPAC Name: 2-chloro-3-methylpyridine
Synonyms: 2-chloro-3-methylpyridine
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: RKVUCIFREKHYTL-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1Cl
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results