missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylpyridine, 98+%, Thermo Scientific™

Product Code. 11359865
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359865 25 g 25g
11349865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359865 Supplier Thermo Scientific Alfa Aesar Supplier No. L19861.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-76-8
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00234177
InChI Key RKVUCIFREKHYTL-UHFFFAOYSA-N
Synonym 2-chloro-3-picoline, 2-chloro 3-methylpyridine, 3-picoline, 2-chloro, pyridine, 2-chloro-3-methyl, 2-chloro-3-methyl-pyridine, pyridine,2-chloro-3-methyl, pubchem2074, 2-chloro-methyl-pyridine, 2-chloro3-methyl-pyridine
PubChem CID 87603
IUPAC Name 2-chloro-3-methylpyridine
SMILES CC1=C(N=CC=C1)Cl

Specifications

Density 1.17
Boiling Point 192°C to 193°C
Flash Point 79°C (174°F)
Refractive Index 1.533
Quantity 25 g
UN Number UN2810
Beilstein 107895
Formula Weight 127.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.