accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1849-27-0

IUPAC Name: 1,4-bis(2-phenylethynyl)benzene
Synonyms: 1,4-bis(2-phenylethynyl)benzene
Molecular Weight (g/mol): 278.35
Molecular Formula: C22H14
InChi Key: FPVSTPLZJLYNMB-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C#CC1=CC=C(C=C1)C#CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results