CAS RN 18585-06-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl (4-Chloro-3-(trifluoromethyl)phenyl)carbamate, TRC
CAS: 18585-06-3 Molecular Formula: C10H9ClF3NO2 Molecular Weight (g/mol): 267.63 Synonym: N-[4-Chloro-3-(trifluoromethyl)phenyl]carbamic Acid Ethyl Ester,[4-Chloro-3-(trifluoromethyl)phenyl]carbamic Acid Ethyl Ester,4-Chloro-3-(trifluoromethyl)carbanilic Acid Ethyl Ester,Sorafenib Impurity 2 SMILES: CCOC(=O)Nc1ccc(Cl)c(c1)C(F)(F)F