missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (4-Chloro-3-(trifluoromethyl)phenyl)carbamate, TRC

Product Code. 30386602
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386602 10 g 10g
30379733 100 mg -
30370706 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386602 Supplier Toronto Research Chemicals Supplier No. TRCE90362510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18585-06-3
Molecular Formula C10H9ClF3NO2
Molecular Weight (g/mol) 267.63
Synonym N-[4-Chloro-3-(trifluoromethyl)phenyl]carbamic Acid Ethyl Ester, [4-Chloro-3-(trifluoromethyl)phenyl]carbamic Acid Ethyl Ester, 4-Chloro-3-(trifluoromethyl)carbanilic Acid Ethyl Ester, Sorafenib Impurity 2
SMILES CCOC(=O)Nc1ccc(Cl)c(c1)C(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H9ClF3NO2/c1-2-17-9(16)15-6-3-4-8(11)7(5-6)10(12,13)14/h3-5H,2H2,1H3,(H,15,16)
Chemical Name or Material Ethyl (4-Chloro-3-(trifluoromethyl)phenyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.