accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClOOHN

CAS RN 18711-13-2

IUPAC Name: 4,7-dichloro-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 4,7-dichloro-1H-indole-2,3-dione
Molecular Weight (g/mol): 216.02
Molecular Formula: C8H3Cl2NO2
InChi Key: NUXYYWOWNFEMNH-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C2=C1NC(=O)C2=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results