accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 187657-92-7

OOBr

CAS RN 187657-92-7

IUPAC Name: 1-(1-benzofuran-3-yl)-2-bromoethan-1-one
Synonyms: 1-(1-benzofuran-3-yl)-2-bromoethanone
Molecular Weight (g/mol): 239.07
Molecular Formula: C10H7BrO2
InChi Key: WVHFTONHSRLBGL-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=COC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results