accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1889-71-0

ClO

CAS RN 1889-71-0

IUPAC Name: 1-(4-chlorophenyl)-2-phenylethan-1-one
Synonyms: 1-(4-chlorophenyl)-2-phenylethanone
Molecular Weight (g/mol): 230.69
Molecular Formula: C14H11ClO
InChi Key: DXVALSKCLLBZEB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results