accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOCl

CAS RN 1892-43-9

IUPAC Name: 2-(4-chlorophenoxy)ethan-1-ol
Synonyms: chlorophetanol ETHYLENEGLYCOLMONO4chlorophenylether
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: GEGSSUSEWOHAFE-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)