missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)ethanol, 98+%

Artikelnummer. 11491848
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11491848 5 g 5g
11401858 25 g 25g
11411858 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 11491848 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A14508.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 1892-43-9 | C8H9ClO2 | 172.608 g/mol

2-(4-Chlorophenoxy)ethanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Chlorophenoxy)ethanol is used as a pharmaceutical intermediate.

Solubility
Soluble in water 3.1 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1892-43-9
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00014036
InChI Key GEGSSUSEWOHAFE-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy ethanol, chlorophetanol, fungisan, ethanol, 2-4-chlorophenoxy, chloro-p-phenoxetol, 2-p-chlorophenoxy ethanol, p-chlorophenyl glycol ether, p-chlorophenyl monoglycol ether, p-chlorfenylmonoglykolether, ethanol, 2-p-chlorophenoxy
PubChem CID 15907
IUPAC Name 2-(4-chlorophenoxy)ethanol
SMILES C1=CC(=CC=C1OCCO)Cl

Tekniske data

Melting Point 30°C
Quantity 5 g
Refractive Index 1.551
UN Number UN2811
Beilstein 1365684
Boiling Point 135°C to 136°C (6 mmHg)
Solubility Information Soluble in water 3.1g/L @ 25°C.
Formula Weight 172.61
Percent Purity ≥98%
Chemical Name or Material 2-(4-Chlorophenoxy)ethanol

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.