accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3H3CH3CCH3

CAS RN 19099-48-0

IUPAC Name: 1-tert-butyl-4-[(4-tert-butylphenyl)methyl]benzene
Synonyms: 1-tert-butyl-4-[(4-tert-butylphenyl)methyl]benzene
Molecular Weight (g/mol): 280.46
Molecular Formula: C21H28
InChi Key: KRMBVBPNWUBFBC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CC2=CC=C(C=C2)C(C)(C)C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results