accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 193090-60-7

NH2BrFFFF

CAS RN 193090-60-7

IUPAC Name: 2-bromo-4-fluoro-5-(trifluoromethyl)aniline
Synonyms: 2-bromo-4-fluoro-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 258.01
Molecular Formula: C7H4BrF4N
InChi Key: ZRMRCSMDONHRKT-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results