missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Product Code. 11336639
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11336639 25 g 25g
11326639 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11336639

Brand: Thermo Scientific Alfa Aesar H26211.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193090-60-7
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.01
MDL Number MFCD04973755
InChI Key ZRMRCSMDONHRKT-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-5-trifluoromethyl aniline, 5-amino-4-bromo-2-fluorobenzotrifluoride, benzenamine, 2-bromo-4-fluoro-5-trifluoromethyl, 2-bromo-4-fluoro-5'-trifluoromethyl aniline, 2-bromo-4-fluoro-5-trifluoromethyl-phenylamine, 2-bromo-4-fluoro-5-trifluoromethyl-benzenamine, acmc-209ewq, ksc496o8r, 2-bromo-4-fluoro-5-trifluoromethyl benzenamine, 2-bromo-4-fluoro-5-trifluoromethyl phenylamine
PubChem CID 24721365
IUPAC Name 2-bromo-4-fluoro-5-(trifluoromethyl)aniline
SMILES NC1=C(Br)C=C(F)C(=C1)C(F)(F)F

Specifications

Melting Point 24°C to 29°C
Quantity 25 g
UN Number UN2811
Formula Weight 258.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluoro-5-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.