accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HN

CAS RN 194-59-2

IUPAC Name: 12-azapentacyclo[11.8.0.0²,¹¹.0³,⁸.0¹⁶,²¹]henicosa-1(13),2(11),3,5,7,9,14,16,18,20-decaene
Synonyms: 7H-dibenzo(C,G)carbazole
Molecular Weight (g/mol): 267.33
Molecular Formula: C20H13N
InChi Key: STJXCDGCXVZHDU-UHFFFAOYSA-N
SMILES: N1C2=C(C3=C1C=CC1=CC=CC=C31)C1=CC=CC=C1C=C2
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results