missing translation for 'onlineSavingsMsg'
Learn More

7H-Dibenzo[c,g]carbazole, TRC

Product Code. 30133590
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133590 100 mg 100mg
30005355 5 mg 5mg
30010820 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133590 Supplier Toronto Research Chemicals Supplier No. TRCD416950100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194-59-2
Molecular Formula C20 H13 N
Molecular Weight (g/mol) 267.32
Synonym 7H-Dibenzo[c,g]carbazole, 3,4:5,6-Dibenzocarbazole, 7-Aza-7H-dibenzo[c,g]fluorene, NSC 87519
SMILES c1ccc2c(c1)ccc3[nH]c4ccc5ccccc5c4c23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H13N/c1-3-7-15-13(5-1)9-11-17-19(15)20-16-8-4-2-6-14(16)10-12-18(20)21-17/h1-12,21H
Formula Weight 267.1048
Chemical Name or Material 7H-Dibenzo[c,g]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.