accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 194787-05-8

CH3OONH4

CAS RN 194787-05-8

IUPAC Name: ammonium acetate
Synonyms: ammonium acetate NH4oac acetic acid, ammonium salt AcONH4
Molecular Weight (g/mol): 77.08
Molecular Formula: C2H7NO2
InChi Key: USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES: [NH4+].CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)