accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHO

CAS RN 194800-16-3

IUPAC Name: 1-pentylcyclopentan-1-ol
Synonyms: 1-pentylcyclopentan-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: LPRXLSZAZXWDMY-UHFFFAOYSA-N
SMILES: CCCCCC1(O)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results