accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBrClO

CAS RN 19652-32-5

IUPAC Name: 3-bromo-5-chloro-2-hydroxybenzaldehyde
Synonyms: 3-bromo-5-chloro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: KNOYZLVIXXBBIB-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Cl)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results