accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNN

CAS RN 19847-12-2

IUPAC Name: pyrazine-2-carbonitrile
Synonyms: cyanopyrazine
Molecular Weight (g/mol): 105.1
Molecular Formula: C5H3N3
InChi Key: PMSVVUSIPKHUMT-UHFFFAOYSA-N
SMILES: N#CC1=CN=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)