missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carbonitrile, Mikromol

Product Code. 30461209
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461209 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461209 Supplier Mikromol Supplier No. MM0385.020100
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pyrazine-2-carbonitrile, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrazinenitrile
CAS 19847-12-2
Analyte or Component Names Pyrazinenitrile
Shipping Condition Room Temperature
Molecular Formula C5 H3 N3
Certifications/Compliance ISO 9001
Synonym Pyrazinonitrile, Pyrazinecarbonitrile, Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitrile, Pyrazinecarbonitrile (7CI,8CI,9CI), Pyrazinonitrile (6CI), 2-Cyanopyrazine, Cyanopyrazine, NSC 166137, NSC 72371, Pyrazinyl cyanide, Pyrazinamide EP Impurity B
InChI Formula InChI=1S/C5H3N3/c6-3-5-4-7-1-2-8-5/h1-2,4H
Solution Type Neat
SMILES N#Cc1cnccn1
IUPAC Name pyrazine-2-carbonitrile
Molecular Weight (g/mol) 105.1
Quantity 100 mg
Formula Weight 105.0327 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.