accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COBrF

CAS RN 198477-89-3

IUPAC Name: 1-(5-bromo-2-fluorophenyl)ethan-1-one
Synonyms: 1-(5-bromo-2-fluorophenyl)ethanone
Molecular Weight (g/mol): 217.04
Molecular Formula: C8H6BrFO
InChi Key: XNRQIHIOKXQSPG-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results