accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 19961-27-4

CH3NHCH3H3C

CAS RN 19961-27-4

IUPAC Name: ethyl(propan-2-yl)amine
Synonyms: ethyl isopropylamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: RIVIDPPYRINTTH-UHFFFAOYSA-N
SMILES: CCNC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results