accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 199682-73-0

IUPAC Name: 5-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde
Synonyms: 3-(4-methoxyphenyl)-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: QSGGFCPKXTULQQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=C(C=O)C=NN1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)