CAS RN 1997296-62-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluconazole N-Oxide, TRC
CAS: 1997296-62-4 Molecular Formula: C13H12F2N6O2 Molecular Weight (g/mol): 322.27 Synonym: Fluconazole mono-N-Oxide SMILES: OC(Cn1cncn1)(Cn2c[n+]([O-])cn2)c3ccc(F)cc3F